Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143255005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325505

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Quantity 0.05mg
Conjugate Unlabeled
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a, κ
Purification Method Affinity Purified
Applications Flow Cytometry
Gene ID (Entrez) 3133
Cross Reactivity Mouse
Clone 28-14-8
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was MHC Class I.
Host Species Mouse
Antigen MHC Class I