Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325501

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Host Species Mouse
Gene ID (Entrez) 3133
Antigen MHC Class I
Immunogen The immunogen for this antibody was MHC Class I.
Storage Requirements Store at 4°C. Do not freeze.
Applications Flow Cytometry
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Primary or Secondary Primary
Quantity 0.1mg
Purification Method Affinity Purified
Clone 28-14-8
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Format Affinity Purified