Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14393405MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1439345

  • / Each
 

Description & Specifications

Specifications

Immunogen The immunogen for this antibody was MHC Class I.
Gene ID (Entrez) 3133
Purification Method Affinity Purified
Cross Reactivity Mouse
Quantity 0.5mg
Format Affinity Purified
Applications Flow Cytometry
Host Species Mouse
Formulation Phosphate buffer pH 7.2
Antigen MHC Class I
Primary or Secondary Primary
Conjugate FITC
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Concentration 0.5mg/mL
Clone 28-14-8
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a, κ