Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 28-14-8, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143934005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14393405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Immunogen The immunogen for this antibody was MHC Class I.
Quantity 0.05mg
Conjugate FITC
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Gene ID (Entrez) 3133
Antigen MHC Class I
Host Species Mouse
Isotype IgG2a, κ
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Clone 28-14-8
Primary or Secondary Primary