Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 14-4-4S, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen MHC Class I
Clone 14-4-4S
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PE MHC class II.
Gene ID (Entrez) 3133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14373601

 
novus biologicals canada
NBP14373601MG
0.1mg Each for $285.00

NBP14373605

 
novus biologicals canada
NBP143736005MG
0.05mg Each for $219.00
Description & Specifications

Specifications

Antigen MHC Class I
Clone 14-4-4S
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PE MHC class II.
Gene ID (Entrez) 3133
Formulation Aqueous Buffer
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry