Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 14-4-4S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143736005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14373605

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Antigen MHC Class I
Formulation Aqueous Buffer
Host Species Mouse
Immunogen The immunogen for this antibody was PE MHC class II.
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3133
Clone 14-4-4S
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL