Please login to your online account to display your discounted pricing

Mouse anti-MHC Class I, Clone: 14-4-4S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14373601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14373601

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Cross Reactivity Rat
Applications Flow Cytometry
Antigen MHC Class I
Gene ID (Entrez) 3133
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Immunogen The immunogen for this antibody was PE MHC class II.
Isotype IgG2a, κ
Formulation Aqueous Buffer
Concentration 0.5mg/mL
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Host Species Mouse
Clone 14-4-4S
Format Affinity Purified