missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: M2-7C10 + M2-9E3 + T311 + HMB45, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Clone M2-7C10 + M2-9E3 + T311 + HMB45
Concentration 1mg/mL
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Gene Accession No. Q16655
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23468101 Novus Biologicals Canada
NBP23468101MG
0.1mg Each for $384.00
NBP23468102 Novus Biologicals Canada
NBP23468102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Melanoma Marker (MART-1 + Tyrosinase + gp100)
1mg/mL
Q16655
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
M2-7C10 + M2-9E3 + T311 + HMB45
Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
2315
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)