Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: M2-7C10 + M2-9E3 + T311 + HMB45, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468102

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 2315
Conjugate Unlabeled
Quantity 0.2mg
Gene Accession No. Q16655
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Cross Reactivity Human
Primary or Secondary Primary
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Host Species Mouse
Reconstitution Protein A purified
Concentration 1mg/mL
Purification Method Purified
Clone M2-7C10 + M2-9E3 + T311 + HMB45
Regulatory Status RUO