Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: M2-7C10 + M2-9E3 + T311 + HMB45, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433705

  • / Each
 

Description & Specifications

Specifications

Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Clone M2-7C10 + M2-9E3 + T311 + HMB45
Concentration 0.2mg/mL
Conjugate Unlabeled