Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: DT101+ BC199 + T311 + HMB45, Novus Biologicals™

$139.00 - $439.00

Specifications

Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Clone DT101+ BC199 + T311 + HMB45
Concentration 0.2mg/mL
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Gene Accession No. Q16655
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23434301

 
novus biologicals canada
NBP23434301ML
0.1mL Each for $139.00

NBP23434305

 
novus biologicals canada
NBP23434305ML
0.5mL Each for $321.00

NBP23460901

 
novus biologicals canada
NBP23460901MG
0.1mg Each for $439.00

NBP23460902

 
novus biologicals canada
NBP23460902MG
0.2mg Each for $439.00
Description & Specifications

Specifications

Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Clone DT101+ BC199 + T311 + HMB45
Concentration 0.2mg/mL
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)