Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: DT101+ BC199 + T311 + HMB45, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460902

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Host Species Mouse
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Format Purified
Clone DT101+ BC199 + T311 + HMB45
Concentration 1mg/mL
Gene Accession No. Q16655
Cross Reactivity Human
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Gene ID (Entrez) 2315
Regulatory Status RUO
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.