Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: A103 + T311 + HMB45, Novus Biologicals™
Manufacturer:  Novus Biologicals Canada NBP23454701MG
 View more versions of this product
Catalog No. NBP23454701
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Melanoma Marker (MART-1 + Tyrosinase + gp100)
A103 + T311 + HMB45
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified
Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
1mg/mL
Purified
Q16655
Recombinant hMART-1 protein (A103); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Purified
RUO
Primary
2315
Human