Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + Tyrosinase + gp100), Clone: A103 + T311 + HMB45, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23433905ML

 View all options for this product


For Research Use Only

Catalog No. NBP23433905


Description & Specifications


Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone A103 + T311 + HMB45
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein (A103); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified