Please login to your online account to display your discounted pricing

Mouse anti-Melanoma Marker (MART-1 + gp100), Clone: DT101+ BC199 + HMB45, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434105

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant hMART-1 protein (DT101 & BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen Melanoma Marker (MART-1 + gp100)
Primary or Secondary Primary
Clone DT101+ BC199 + HMB45