Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM555, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23297910ML

 View all options for this product


For Research Use Only

Catalog No. NBP23297910


Description & Specifications


Antigen Melan-A/MART-1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM555
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified