Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM555, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297901

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Melan-A/MART-1
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Clone SPM555
Format Purified
Gene Accession No. Q16655
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Primary or Secondary Primary