Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM555, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479702

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.2mg
Test Specificity This antibody recognizes a protein doublet of 20-22kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A.
Clone SPM555
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Concentration 1mg/mL