missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM555, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23479701MG

 View more versions of this product

Catalog No. NBP23479701


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Melan-A/MART-1
SPM555
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1, κ
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes a protein doublet of 20-22kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A.
Canine,Human,Murine,Rat
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
1mg/mL
Purified
Q16655
Recombinant hMART-1 protein
Purified
RUO
Primary
2315
Protein A purified