Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM540, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297701

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant hMART-1 protein
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Clone SPM540
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL