missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM540, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23478602MG

 View more versions of this product

Catalog No. NBP23478602


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Melan-A/MART-1
SPM540
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG2b, κ
0.2mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
1mg/mL
Purified
Q16655
Recombinant hMART-1 protein
Purified
RUO
Primary
2315
Human,Murine,Rat