Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM342, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297610

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Host Species Mouse
Quantity 1.0mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Conjugate Unlabeled
Cross Reactivity Human
Antigen Melan-A/MART-1
Formulation PBS with 0.05% BSA.
Immunogen Recombinant hMART-1 protein
Isotype IgG2b, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Does not react to Mouse and Rat.
Reconstitution Protein A purified
Clone SPM342
Concentration 0.2mg/mL
Gene Accession No. Q16655