missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: SPM342, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Melan-A/MART-1
Clone SPM342
Concentration 1mg/mL
Immunogen Recombinant hMART-1 protein
Gene Accession No. Q16655
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23476501 Novus Biologicals Canada
NBP23476501MG
0.1mg Each for $384.00
NBP23476502 Novus Biologicals Canada
NBP23476502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Melan-A/MART-1
1mg/mL
Q16655
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM342
Recombinant hMART-1 protein
2315
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot