Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-9E3, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen Melan-A/MART-1
Clone M2-9E3
Concentration 0.2mg/mL
Immunogen Recombinant human Melan-A protein was used as immunogen to generate the Melan-A antibody.
Gene Accession No. Q16655
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21519801

 
novus biologicals canada
NBP21519801MG
0.1mg Each for $329.00

NBP21519802

 
novus biologicals canada
NBP21519802MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen Melan-A/MART-1
Clone M2-9E3
Concentration 0.2mg/mL
Immunogen Recombinant human Melan-A protein was used as immunogen to generate the Melan-A antibody.
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting