Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-9E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519802

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Antigen Melan-A/MART-1
Format Purified
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This antibody recognizes a protein doublet of 20-22kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This MAb labels melanomas and other tumors showing melanocytic differentiation. It is also a useful positive-marker for angiomyolipomas. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. Q16655
Quantity 0.2mg
Immunogen Recombinant human Melan-A protein was used as immunogen to generate the Melan-A antibody.
Regulatory Status RUO
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation PBS with 0.05% BSA. Protein G purified.
Concentration 0.2mg/mL
Isotype IgG2b, κ
Primary or Secondary Primary
Gene ID (Entrez) 2315
Clone M2-9E3