Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-7C10 + M2-9E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424505

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Formulation PBS with 0.05% BSA.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2b
Regulatory Status RUO
Clone M2-7C10 + M2-9E3
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2315
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL