Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-7C10 + M2-9E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424501

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Quantity 0.1mL
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene ID (Entrez) 2315
Formulation PBS with 0.05% BSA.
Isotype IgG2b
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Concentration 0.2mg/mL
Gene Accession No. Q16655
Monoclonal or Polyclonal Monoclonal
Clone M2-7C10 + M2-9E3
Host Species Mouse
Antigen Melan-A/MART-1
Purification Method Purified
Reconstitution Protein A purified
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)