Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-7C10 + M2-9E3, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Melan-A/MART-1
Clone M2-7C10 + M2-9E3
Concentration 1mg/mL
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Gene Accession No. Q16655
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23468001

 
novus biologicals canada
NBP23468001MG
0.1mg Each for $384.00

NBP23468002

 
novus biologicals canada
NBP23468002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Melan-A/MART-1
Clone M2-7C10 + M2-9E3
Concentration 1mg/mL
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2b
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot