Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: M2-7C10 + M2-9E3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468001

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Isotype IgG2b
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone M2-7C10 + M2-9E3
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Melan-A/MART-1
Format Purified
Gene Accession No. Q16655
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Conjugate Unlabeled