Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: DT101+ BC199, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424605

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Antigen Melan-A/MART-1
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Isotype IgG2b
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 2315
Clone DT101+ BC199
Gene Accession No. Q16655
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat