Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: DT101+ BC199, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460701

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16655
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Isotype IgG2b
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Clone DT101+ BC199
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Reconstitution Protein A purified