Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: DT101+ BC199, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460702

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone DT101+ BC199
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16655
Host Species Murine
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3)
Isotype IgG2b
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Target Species Human
Target Species Murine
Target Species Rat