Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: A103 + M2-7C10 + M2-9E3, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Melan-A/MART-1
Clone A103 + M2-7C10 + M2-9E3
Concentration 0.2mg/mL
Immunogen Recombinant hMART-1 protein (A103; M2-7C10; M2-9E3)
Gene Accession No. Q16655
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23424405

 
novus biologicals canada
NBP23424405ML
0.5mL Each for $321.00

NBP23424401

 
novus biologicals canada
NBP23424401ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Melan-A/MART-1
Clone A103 + M2-7C10 + M2-9E3
Concentration 0.2mg/mL
Immunogen Recombinant hMART-1 protein (A103; M2-7C10; M2-9E3)
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting