missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: A103 + M2-7C10 + M2-9E3, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23454601MG

 View more versions of this product

Catalog No. NBP23454601


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Melan-A/MART-1
A103 + M2-7C10 + M2-9E3
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
1mg/mL
Purified
Q16655
Recombinant hMART-1 protein (A103; M2-7C10; M2-9E3)
Purified
RUO
Primary
2315
Canine,Human,Murine,Rat