Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: A103, Azide Free, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23454502MG

 View all options for this product


For Research Use Only

Catalog No. NBP23454502


Description & Specifications


Antigen Melan-A/MART-1
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone A103
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified