Please login to your online account to display your discounted pricing

Mouse anti-Melan-A/MART-1, Clone: A103, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454501

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Format Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315
Reconstitution Protein A purified
Clone A103
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat