Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: P1H12, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen MCAM/CD146
Clone P1H12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD146.
Gene ID (Entrez) 4162
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14334601

 
novus biologicals canada
NBP14334601MG
0.1mg Each for $285.00

NBP14334603

 
novus biologicals canada
NBP1433460025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen MCAM/CD146
Clone P1H12
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD146.
Gene ID (Entrez) 4162
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting