Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: P1H12, FITC, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen MCAM/CD146
Clone P1H12
Immunogen The immunogen for this antibody was CD146.
Gene ID (Entrez) 4162
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200069

 
novus biologicals canada
NBP200069100TESTS
100 Tests Each for $329.00

NBP20006925

 
novus biologicals canada
NBP20006925TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen MCAM/CD146
Clone P1H12
Immunogen The immunogen for this antibody was CD146.
Gene ID (Entrez) 4162
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry