Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: OJ79c, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen MCAM/CD146
Clone OJ79c
Concentration 0.2mg/mL
Immunogen Recombinant human MUC18 protein
Gene Accession No. P43121
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23427501

 
novus biologicals canada
NBP23427501ML
0.1mL Each for $139.00

NBP23427505

 
novus biologicals canada
NBP23427505ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen MCAM/CD146
Clone OJ79c
Concentration 0.2mg/mL
Immunogen Recombinant human MUC18 protein
Gene Accession No. P43121
Gene ID (Entrez) 4162
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting