Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: OJ79c, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427505

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human MUC18 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen MCAM/CD146
Format Purified
Gene Accession No. P43121
Host Species Mouse
Regulatory Status RUO
Gene ID (Entrez) 4162
Reconstitution Protein A purified
Clone OJ79c
Concentration 0.2mg/mL
Cross Reactivity Human