Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: OJ79c, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23427501ML

 View all options for this product


For Research Use Only

Catalog No. NBP23427501


Description & Specifications


Antigen MCAM/CD146
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone OJ79c
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P43121
Host Species Mouse
Immunogen Recombinant human MUC18 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4162
Reconstitution Protein A purified