Please login to your online account to display your discounted pricing

Mouse anti-MCAM/CD146, Clone: OJ79c, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470101

  • / Each
 

Description & Specifications

Specifications

Antigen MCAM/CD146
Format Purified
Gene Accession No. P43121
Immunogen Recombinant human MUC18 protein
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4162
Conjugate Unlabeled
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone OJ79c
Reconstitution Protein A purified
Concentration 1mg/mL