Please login to your online account to display your discounted pricing

Mouse anti-MALT1, Clone: SPM578, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481102

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Antigen MALT1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Human MALT1 recombinant fragment (aa701-808)
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10892
Clone SPM578
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. Q9UDY8
Purification Method Purified
Regulatory Status RUO
Reconstitution Protein A purified
Concentration 1mg/mL