Please login to your online account to display your discounted pricing

Mouse anti-MALT1, Clone: 50, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen MALT1
Clone 50
Immunogen The immunogen for this antibody was MALT1.
Gene ID (Entrez) 10892
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14312001

 
novus biologicals canada
NBP14312001MG
0.1mg Each for $362.00

NBP14312003

 
novus biologicals canada
NBP1431200025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen MALT1
Clone 50
Immunogen The immunogen for this antibody was MALT1.
Gene ID (Entrez) 10892
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting