Please login to your online account to display your discounted pricing

Mouse anti-MALT1, Clone: 50, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431200025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14312003

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Conjugate Unlabeled
Isotype IgG1
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.025mg
Immunogen The immunogen for this antibody was MALT1.
Regulatory Status RUO
Format Affinity Purified
Purification Method Affinity Purified
Antigen MALT1
Gene ID (Entrez) 10892
Host Species Mouse
Primary or Secondary Primary
Clone 50