Please login to your online account to display your discounted pricing

Mouse anti-MAGE 1, Clone: SPM282, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287101

  • / Each
 

Description & Specifications

Specifications

Antigen MAGE 1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P43355
Immunogen Human MAGE-A1 full length recombinant protein
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4100
Reconstitution Protein A purified
Clone SPM282
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Host Species Mouse
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL