Please login to your online account to display your discounted pricing

Mouse anti-MAGE 1, Clone: SPM282, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287110

  • / Each
 

Description & Specifications

Specifications

Antigen MAGE 1
Format Purified
Gene Accession No. P43355
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human MAGE-A1 full length recombinant protein
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4100
Reconstitution Protein A purified
Clone SPM282
Concentration 0.2mg/mL