Please login to your online account to display your discounted pricing

Mouse anti-MAGE 1, Clone: SPM282, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MAGE 1
Clone SPM282
Concentration 1mg/mL
Immunogen Human MAGE-A1 full length recombinant protein
Gene Accession No. P43355
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23475401

 
novus biologicals canada
NBP23475401MG
0.1mg Each for $384.00

NBP23475402

 
novus biologicals canada
NBP23475402MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen MAGE 1
Clone SPM282
Concentration 1mg/mL
Immunogen Human MAGE-A1 full length recombinant protein
Gene Accession No. P43355
Gene ID (Entrez) 4100
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting