Please login to your online account to display your discounted pricing

Mouse anti-MAGE 1, Clone: SPM282, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475402

  • / Each
 

Description & Specifications

Specifications

Antigen MAGE 1
Format Purified
Host Species Mouse
Concentration 1mg/mL
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P43355
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a protein of 42-46kDa, identified as MAGE-1. This MAb does not cross-react with MAGE-2, -3, -4, -6 -9, -10, -or -12 protein.
Reconstitution Protein A purified
Clone SPM282
Immunogen Human MAGE-A1 full length recombinant protein
Isotype IgG1, κ
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 4100
Conjugate Unlabeled