Please login to your online account to display your discounted pricing

Mouse anti-MAGE 1, Clone: SPM282, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Immunogen Human MAGE-A1 full length recombinant protein
Format Purified
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 4100
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Host Species Mouse
Gene Accession No. P43355
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a protein of 42-46kDa, identified as MAGE-1. This MAb does not cross-react with MAGE-2, -3, -4, -6 -9, -10, -or -12 protein.
Clone SPM282
Antigen MAGE 1
Primary or Secondary Primary